Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bodipy 558/568 C12 | 158757-84-7 | 98.4% | 472.40 g/mol | C25H31BF2N2O2S | 5 MG
SDP

Catalog No. 5000222848
Encompass_Preferred

A lipophilic BODIPY fluorescent dye for selective staining of neutral lipid droplets in live or fixed cells, providing bright fluorescence with excitation/emission maxima at 558/568 nm and high chemical purity for imaging applications.

  • Selective lipid droplet staining in live and fixed cells.
  • Excitation/emission maxima 558/568 nm.
  • High purity (98.4%) for consistent imaging results.
  • Soluble in DMSO (≈125 mg/mL); sonication may be required.
  • Stable when stored protected from light and moisture; solutions stable at -80°C up to 6 months.
  • Suitable for fluorescence microscopy and lipid trafficking studies.

Catalog No. 50-002-22848 Supplier Medchemexpress LLC Supplier No. HY1382265MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

A lipophilic BODIPY fluorescent dye for selective staining of neutral lipid droplets in live or fixed cells, providing bright fluorescence with excitation/emission maxima at 558/568 nm and high chemical purity for imaging applications.

  • Selective lipid droplet staining in live and fixed cells.
  • Excitation/emission maxima 558/568 nm.
  • High purity (98.4%) for consistent imaging results.
  • Soluble in DMSO (≈125 mg/mL); sonication may be required.
  • Stable when stored protected from light and moisture; solutions stable at -80°C up to 6 months.
  • Suitable for fluorescence microscopy and lipid trafficking studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.