Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BODIPY 558/568 C12 | 158757-84-7 | 98.4% | C25H30BF2N2O2S • H | 10MG
SDP

Catalog No. 5000260633
Encompass_Preferred

BODIPY 558/568 C12 is a membrane-permeant, fatty-acid-conjugated BODIPY fluorescent dye that selectively labels neutral lipid droplets in live or fixed cells. It provides bright, photostable orange-red fluorescence and is suitable for fluorescence microscopy studies of lipid localization and dynamics.

  • Excitation/emission maxima 558 nm / 568 nm.
  • Membrane-permeant small-molecule probe for lipid droplets.
  • Bright, photostable orange-red fluorescence for imaging.
  • Suitable for live-cell and fixed-cell labeling.
  • Reported molecular weight ~472.4 g·mol⁻¹.
  • Purity reported ~98.4% (sources vary).

Catalog No. 50-002-60633 Supplier Medchemexpress LLC Supplier No. HY13822610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BODIPY 558/568 C12 is a membrane-permeant, fatty-acid-conjugated BODIPY fluorescent dye that selectively labels neutral lipid droplets in live or fixed cells. It provides bright, photostable orange-red fluorescence and is suitable for fluorescence microscopy studies of lipid localization and dynamics.

  • Excitation/emission maxima 558 nm / 568 nm.
  • Membrane-permeant small-molecule probe for lipid droplets.
  • Bright, photostable orange-red fluorescence for imaging.
  • Suitable for live-cell and fixed-cell labeling.
  • Reported molecular weight ~472.4 g·mol⁻¹.
  • Purity reported ~98.4% (sources vary).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.