Learn More
Medchemexpress LLC Bodipy 558/568 C12 | 158757-84-7 | 98.42% | 472.40 | 1 MG
SDP
Catalog No. 5000347763
Encompass_Preferred
BODIPY 558/568 C12 is a small molecule BODIPY dye known for its strong ultraviolet absorption, sharp fluorescence peaks, and high quantum yield. It maintains stability across various physiological conditions, being relatively insensitive to environmental polarity and pH. Its structural asymmetry allows for diverse structural applications.
  • Strong ultraviolet absorption, sharp fluorescence peaks, and high quantum yield
  • Relatively insensitive to environmental polarity and pH
  • Maintains stability under various physiological conditions
  • Structural asymmetry offers diverse structural products
  • Can readily pass through cell membranes
  • Localizes neutral lipids within cells to specifically stain lipid droplets
  • Suitable for labeling both live and fixed cells
  • Maximum excitation/emission wavelength is 558/568 nm
Catalog No. 50-003-47763 Supplier Medchemexpress LLC Supplier No. HY1382261MG
Add to Cart
Add to Cart
BODIPY 558/568 C12 is a small molecule BODIPY dye known for its strong ultraviolet absorption, sharp fluorescence peaks, and high quantum yield. It maintains stability across various physiological conditions, being relatively insensitive to environmental polarity and pH. Its structural asymmetry allows for diverse structural applications.
  • Strong ultraviolet absorption, sharp fluorescence peaks, and high quantum yield
  • Relatively insensitive to environmental polarity and pH
  • Maintains stability under various physiological conditions
  • Structural asymmetry offers diverse structural products
  • Can readily pass through cell membranes
  • Localizes neutral lipids within cells to specifically stain lipid droplets
  • Suitable for labeling both live and fixed cells
  • Maximum excitation/emission wavelength is 558/568 nm