Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BODIPY 576/589 | 150173-78-7 | 96.18% | 329.11 | 1 ML
SDP

Catalog No. 5000428172
Encompass_Preferred

BODIPY 576/589 is a long-wavelength biological labeled dye that contains a free COOH-group, with Em/Ex = 589/576 nm. It is for research use only and not sold to patients.

  • Long wavelength biological labeled dye.
  • Contains a free COOH-group.
  • Emission (Em) at 589 nm.
  • Excitation (Ex) at 576 nm.

Catalog No. 50-004-28172 Supplier Medchemexpress LLC Supplier No. HYD111810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BODIPY 576/589 is a long-wavelength biological labeled dye that contains a free COOH-group, with Em/Ex = 589/576 nm. It is for research use only and not sold to patients.

  • Long wavelength biological labeled dye.
  • Contains a free COOH-group.
  • Emission (Em) at 589 nm.
  • Excitation (Ex) at 576 nm.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.