Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BODIPY 576/589 | 150173-78-7 | 96.2% | 329.11 | 5 MG
SDP

Catalog No. 5000393291
Encompass_Preferred

BODIPY 576/589 is a long wavelength biological labeled dye for research use. It contains a free COOH-group and features emission (Em) at 589 nm and excitation (Ex) at 576 nm. This dye is suitable for various biological conjugation and protein labeling applications.

  • Long wavelength biological labeled dye
  • Contains a free COOH-group
  • Emission wavelength at 589 nm
  • Excitation wavelength at 576 nm
  • Suitable for protein labeling protocols
  • Stable under recommended storage conditions

Catalog No. 50-003-93291 Supplier Medchemexpress LLC Supplier No. 5MGHYD1118
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BODIPY 576/589 is a long wavelength biological labeled dye for research use. It contains a free COOH-group and features emission (Em) at 589 nm and excitation (Ex) at 576 nm. This dye is suitable for various biological conjugation and protein labeling applications.

  • Long wavelength biological labeled dye
  • Contains a free COOH-group
  • Emission wavelength at 589 nm
  • Excitation wavelength at 576 nm
  • Suitable for protein labeling protocols
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.