Learn More
Medchemexpress LLC Bodipy 581/591 C11 | 217075-36-0 | >95.0% | 504.42 | C30H35BF2N2O2 | 10 MG
SDP
Catalog No. 5000297215
Encompass_Preferred
BODIPY 581/591 C11 is an oxidation-sensitive fluorescent probe used to detect lipid peroxidation and oxidative stress in living cells. It exhibits a ratiometric fluorescence shift from red to green upon oxidation, allowing quantification of lipid oxidation and detection of ferroptosis in cell-based assays.
  • Oxidation-sensitive ratiometric probe for lipid peroxidation monitoring.
  • Dual excitation/emission enables red-to-green shift detection in cells.
  • Small-molecule dye form suitable for cell-permeable assays.
  • Available in small milligram quantities for assay development and research.
  • Stable when stored lyophilized at low temperature for typical laboratory use.
Catalog No. 50-002-97215 Supplier Medchemexpress LLC Supplier No. HYD130110MG
BODIPY 581/591 C11 is an oxidation-sensitive fluorescent probe used to detect lipid peroxidation and oxidative stress in living cells. It exhibits a ratiometric fluorescence shift from red to green upon oxidation, allowing quantification of lipid oxidation and detection of ferroptosis in cell-based assays.
  • Oxidation-sensitive ratiometric probe for lipid peroxidation monitoring.
  • Dual excitation/emission enables red-to-green shift detection in cells.
  • Small-molecule dye form suitable for cell-permeable assays.
  • Available in small milligram quantities for assay development and research.
  • Stable when stored lyophilized at low temperature for typical laboratory use.