Learn More
Medchemexpress LLC BODIPY-581/591 NHS ester | 150152-70-8 | 98.67% | 489.28 | 5 MG
SDP
Catalog No. 5000429997
Encompass_Preferred
BODIPY-581/591 NHS ester is a bright, red fluorescent dye used for the labeling of amine containing biomolecules. It exhibits excitation at 581 nm and emission at 591 nm.
  • It is a bright, red fluorescent dye.
  • It has an excitation wavelength of 581 nm.
  • It has an emission wavelength of 591 nm.
  • It can be used for labeling biomolecules that contain amine groups.
Catalog No. 50-004-29997 Supplier Medchemexpress LLC Supplier No. HYD16915MG
BODIPY-581/591 NHS ester is a bright, red fluorescent dye used for the labeling of amine containing biomolecules. It exhibits excitation at 581 nm and emission at 591 nm.
  • It is a bright, red fluorescent dye.
  • It has an excitation wavelength of 581 nm.
  • It has an emission wavelength of 591 nm.
  • It can be used for labeling biomolecules that contain amine groups.