Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bodipy 630/650x | 380367-48-6 | MFCD30182301 | 99.2% | 660.50 | C33H31BF2N4O6S | 5 MG
SDP

Catalog No. 5000263536
Encompass_Preferred

BODIPY 630/650X is a red/far-red fluorescent conjugate of the adenosine receptor ligand N-ethylcarboxamido-adenosine (NECA). It provides excitation and emission maxima at 630 nm and 650 nm, respectively, and is formulated as a solid dye for fluorescence imaging, labeling, and detection applications.

  • Excitation/emission maxima 630 nm / 650 nm
  • High purity (99.24%)
  • Solid, dark purple to reddish brown appearance
  • Chemical formula C33H31BF2N4O6S; molecular weight 660.50
  • Storage: store sealed at 4°C, protected from light and moisture
  • In solvent: -80°C up to 6 months; -20°C up to 1 month
  • Suitable for fluorescent labeling and imaging of adenosine receptor targets

Catalog No. 50-002-63536 Supplier Medchemexpress LLC Supplier No. HY1102135MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BODIPY 630/650X is a red/far-red fluorescent conjugate of the adenosine receptor ligand N-ethylcarboxamido-adenosine (NECA). It provides excitation and emission maxima at 630 nm and 650 nm, respectively, and is formulated as a solid dye for fluorescence imaging, labeling, and detection applications.

  • Excitation/emission maxima 630 nm / 650 nm
  • High purity (99.24%)
  • Solid, dark purple to reddish brown appearance
  • Chemical formula C33H31BF2N4O6S; molecular weight 660.50
  • Storage: store sealed at 4°C, protected from light and moisture
  • In solvent: -80°C up to 6 months; -20°C up to 1 month
  • Suitable for fluorescent labeling and imaging of adenosine receptor targets

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.