Learn More
Cayman Chemical BODIPY 630/650X succinimide ester, Purity: ≥90%, CAS: 380367-48-6, 1 mg
SDP
Catalog No. 502940669
Encompass_Preferred
An amine-reactive fluorescent probe; has been used, linked to adenosine A1 receptor ligands, as a fluorescent probe to quantify ligand-receptor binding; ex/em = 630/650 nm, respectively
Catalog No. 50-294-0669 Supplier Cayman Chemical Supplier No. 277581 MG
An amine-reactive fluorescent probe; has been used, linked to adenosine A1 receptor ligands, as a fluorescent probe to quantify ligand-receptor binding; ex/em = 630/650 nm, respectively