Learn More
Medchemexpress LLC Bodipy-C12 | 1029138-21-3 | 99.8% | 452.39 | C27H35BF2N2O | 10 MG
SDP
Catalog No. 5000298844
Encompass_Preferred
BODIPY-C12 is a boron-dipyrromethene fluorescent probe bearing a C12 lipophilic tail. The hydrophobic dye is sensitive to its local environment and is used in fluorescence-based assays to quantify microviscosity, critical micelle concentration, and critical micelle temperature.
  • Hydrophobic BODIPY dye with a C12 lipophilic tail.
  • Excitation 490 nm and emission 512 nm for green fluorescence.
  • High purity (~99.8%) suitable for sensitive fluorescence measurements.
  • Available in small laboratory quantities, including 10 MG packages.
  • Store solid at 4°C protected from light; in solvent store -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-98844 Supplier Medchemexpress LLC Supplier No. HYD155310MG
BODIPY-C12 is a boron-dipyrromethene fluorescent probe bearing a C12 lipophilic tail. The hydrophobic dye is sensitive to its local environment and is used in fluorescence-based assays to quantify microviscosity, critical micelle concentration, and critical micelle temperature.
  • Hydrophobic BODIPY dye with a C12 lipophilic tail.
  • Excitation 490 nm and emission 512 nm for green fluorescence.
  • High purity (~99.8%) suitable for sensitive fluorescence measurements.
  • Available in small laboratory quantities, including 10 MG packages.
  • Store solid at 4°C protected from light; in solvent store -80°C (6 months) or -20°C (1 month).