Learn More
Kyfora Bio BODIPY-Cholesterol 1 MG
Catalog No. 502559792
Encompass_Preferred
BODIPY-Cholesterol is a fluorescently labeled cholesterol analog featuring the BODIPY fluorophore, ideal for tracking and studying cholesterol dynamics within lipid nanoparticles (LNPs). Cholesterol is a critical component in LNP formulations, providing essential structural integrity and stability to lipid bilayers. Conjugation with BODIPY enables visualization of cellular uptake and intracellular trafficking of LNPs, facilitating real-time monitoring in various research applications. BODIPY-Cholesterol exhibits an excitation/emission profile of 480/508 nm, making it compatible with a wide range of fluorescence detection systems.Applications: LNP formulation, cellular uptake monitoring
Catalog No. 50-255-9792 Supplier Kyfora Bio Supplier No. 145261
BODIPY-Cholesterol is a fluorescently labeled cholesterol analog featuring the BODIPY fluorophore, ideal for tracking and studying cholesterol dynamics within lipid nanoparticles (LNPs). Cholesterol is a critical component in LNP formulations, providing essential structural integrity and stability to lipid bilayers. Conjugation with BODIPY enables visualization of cellular uptake and intracellular trafficking of LNPs, facilitating real-time monitoring in various research applications. BODIPY-Cholesterol exhibits an excitation/emission profile of 480/508 nm, making it compatible with a wide range of fluorescence detection systems.Applications: LNP formulation, cellular uptake monitoring