Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BODIPY™ FL (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionic Acid)

Catalog No. D2183
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2183 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2183 Supplier Invitrogen™ Supplier No. D2183
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

BODIPY™ FL (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionic Acid)

The amine-reactive BODIPY™ FL propionic acid can be used to create green-fluorescent bioconjugates following carbodiimide-activation of the carboxylic acid on the dye and reaction with amines. The electronically neutral BODIPY™ FL dye has the most fluorescein-like spectra of the green-fluorescent BODIPY™ dyes.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type FL Propionic Acid
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Carboxylic Acid
Label or Dye BODIPY™ FL
Label Type BODIPY Dyes
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.