Learn More
Medchemexpress LLC BODIPY FL alkyne | 25MG
SDP
Catalog No. 5000229113
Encompass_Preferred
BODIPY FL alkyne (BOD FL alkyne) is a green-fluorescent dye (Ex 503 nm Em 512 nm) a bright and photostable fluorophore which emits in fluorescein (FAM) channel BODIPY FL alkyne can be conjugated with a number of azide-containing molecules by copper-catalyzed Click Chemistry
Catalog No. 50-002-29113 Supplier Medchemexpress LLC Supplier No. HY11435425MG
BODIPY FL alkyne (BOD FL alkyne) is a green-fluorescent dye (Ex 503 nm Em 512 nm) a bright and photostable fluorophore which emits in fluorescein (FAM) channel BODIPY FL alkyne can be conjugated with a number of azide-containing molecules by copper-catalyzed Click Chemistry