Learn More
Medchemexpress LLC Bodipy FL C5-ceramide | 133867-53-5 | 98.9% | 601.62 | C34H54BF2N3O3 | 10 MG
SDP
Catalog No. 5000297850
Encompass_Preferred
BODIPY FL C5-Ceramide is a BODIPY-labeled ceramide derivative used as a Golgi-specific green fluorescent dye for fluorescence microscopy. It is synthesized in the endoplasmic reticulum and trafficked to the Golgi apparatus, enabling selective labeling of Golgi membranes in live and fixed cells.
  • Golgi-specific fluorescent labeling for microscopy.
  • Bright green fluorescence with excitation/emission at 505/511 nm.
  • High purity suitable for imaging applications.
  • Solid, brown-to-red appearance; stable when stored protected from light at -20 °C.
  • Supplied in small-mass quantities suitable for laboratory imaging workflows.
Catalog No. 50-002-97850 Supplier Medchemexpress LLC Supplier No. HYD161210MG
BODIPY FL C5-Ceramide is a BODIPY-labeled ceramide derivative used as a Golgi-specific green fluorescent dye for fluorescence microscopy. It is synthesized in the endoplasmic reticulum and trafficked to the Golgi apparatus, enabling selective labeling of Golgi membranes in live and fixed cells.
  • Golgi-specific fluorescent labeling for microscopy.
  • Bright green fluorescence with excitation/emission at 505/511 nm.
  • High purity suitable for imaging applications.
  • Solid, brown-to-red appearance; stable when stored protected from light at -20 °C.
  • Supplied in small-mass quantities suitable for laboratory imaging workflows.