Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bodipy FL C5-HPC | 161106-88-3 | 99.4% | 797.76 g/mol | C40H67BF2N3O8P | 10 MG
SDP

Catalog No. 5000296695
Encompass_Preferred

BODIPY FL C5-HPC is a lipophilic green fluorescent dye with excitation/emission maxima near 500/510 nm, intended for labeling membranes and lipophilic structures in fluorescence microscopy and flow cytometry.

  • Excitation/emission: 500/510 nm for green fluorescence.
  • Lipophilic dye suitable for membrane and lipid labeling.
  • High purity: 99.4% (HPLC).
  • Molecular weight: 797.76 g/mol; CAS: 161106-88-3.
  • Supplied in low-mass vials suitable for laboratory use and custom assays.

Catalog No. 50-002-96695 Supplier Medchemexpress LLC Supplier No. HYD161310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BODIPY FL C5-HPC is a lipophilic green fluorescent dye with excitation/emission maxima near 500/510 nm, intended for labeling membranes and lipophilic structures in fluorescence microscopy and flow cytometry.

  • Excitation/emission: 500/510 nm for green fluorescence.
  • Lipophilic dye suitable for membrane and lipid labeling.
  • High purity: 99.4% (HPLC).
  • Molecular weight: 797.76 g/mol; CAS: 161106-88-3.
  • Supplied in low-mass vials suitable for laboratory use and custom assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.