Learn More
Medchemexpress LLC BODIPY FL Ethylamine | 1229571-00-9 | 98.1% | 291.15 | 100 MG
SDP
Catalog No. 5000429999
Encompass_Preferred
BODIPY FL Ethylamine is a fluorescent dye that reversibly reacts with aldehyde/ketone to yield a Schiff base. It can be reduced to form a stable amine derivative using sodium borohydride or sodium cyanoborohydride. It is for research use only and not sold to patients.
  • Fluorescent dye
  • Reversibly reacts with aldehyde/ketone
  • Yields a Schiff base
  • Can be reduced to form a stable amine derivative
  • For research use only
Catalog No. 50-004-29999 Supplier Medchemexpress LLC Supplier No. HYD1604100MG
BODIPY FL Ethylamine is a fluorescent dye that reversibly reacts with aldehyde/ketone to yield a Schiff base. It can be reduced to form a stable amine derivative using sodium borohydride or sodium cyanoborohydride. It is for research use only and not sold to patients.
  • Fluorescent dye
  • Reversibly reacts with aldehyde/ketone
  • Yields a Schiff base
  • Can be reduced to form a stable amine derivative
  • For research use only