Learn More
Medchemexpress LLC BODIPY FL hydrazide 10mg | 178388-71-1 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11435110MG
Encompass_Preferred
BODIPY FL hydrazide is a green-fluorescent hydrazide reagent that reacts with aldehyde and ketone groups on polysaccharides and glycoproteins to form a reversible Schiff base that can be reduced to a stable linkage for detection and imaging. Its excitation maximum is about 495 nm and its emission maximum is about 516 nm.
  • Reactive toward aldehyde and ketone groups
  • Forms a reversible Schiff base convertible to a stable linkage by reduction
  • Green fluorescence with excitation ~495 nm and emission ~516 nm
  • Suitable for fluorescent labeling of carbohydrates and glycoproteins
Catalog No. 50-002-29114