Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BODIPY FL hydrazide 25mg | 178388-71-1 | 25 MG
SDP

Catalog No. 5000229742
Encompass_Preferred

BODIPY FL hydrazide is a green-fluorescent hydrazide reagent used to label aldehyde and ketone groups on polysaccharides and glycoproteins; it forms a reversible Schiff base that can be reduced to a stable linkage.

  • Green-fluorescent hydrazide suitable for labeling aldehyde and ketone groups.
  • Excitation 495 nm, emission 516 nm, compatible with common FITC/GFP filter sets.
  • High purity (97.4%) and solid brown to red appearance.
  • Soluble in DMSO (125 mg/mL) with ultrasonic assistance; use newly opened DMSO.
  • Store protected from light; solid at 4°C; in solution store -80°C (6 months) or -20°C (1 month).
  • Forms a Schiff base that can be stabilized by reduction (for example, sodium borohydride).

Catalog No. 50-002-29742 Supplier Medchemexpress LLC Supplier No. HY11435125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BODIPY FL hydrazide is a green-fluorescent hydrazide reagent used to label aldehyde and ketone groups on polysaccharides and glycoproteins; it forms a reversible Schiff base that can be reduced to a stable linkage.

  • Green-fluorescent hydrazide suitable for labeling aldehyde and ketone groups.
  • Excitation 495 nm, emission 516 nm, compatible with common FITC/GFP filter sets.
  • High purity (97.4%) and solid brown to red appearance.
  • Soluble in DMSO (125 mg/mL) with ultrasonic assistance; use newly opened DMSO.
  • Store protected from light; solid at 4°C; in solution store -80°C (6 months) or -20°C (1 month).
  • Forms a Schiff base that can be stabilized by reduction (for example, sodium borohydride).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.