Learn More
Invitrogen™ BODIPY™ FL L-Cystine
Description
We have attached two BODIPY FL fluorophores to the amino groups of the disulfide-containing amino acid cystine to create this reagent for reversible thiol-specific labeling of thiolated oligonucleotides, proteins and cells. BODIPY FL L-cystine is virtually nonfluorescent due to interactions between the two fluorophores; however, thiol-specific exchange with thiolated biomolecules occurs to form mixed disulfides, resulting in green fluorescence.
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | FL L-Cystine |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Thiol |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Thiol |
| Label or Dye | BODIPY™ FL |
| Label Type | BODIPY Dyes |
| Product Line | BODIPY |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.