Learn More
Medchemexpress LLC BODIPY-FL NHS ester | 146616-66-2 | 99.8% | 389.16 | 1 MG
SDP
Catalog No. 5000344140
Encompass_Preferred
BODIPY FL NHS ester (BODIPY FL, SE) is a cell membrane-penetrable amine-reactive fluorescent probe. The maximum excitation/emission wavelength of the BODIPY-FL NHS ester are 502/511 nm, respectively. It has high stability and is insensitive to the polarity, pH and type of solvent, and can maintain stable fluorescence properties under different environmental conditions. It can be used for the synthesis of protease substrates, live cell imaging, protein labeling and immunoassay.
  • Cell membrane-penetrable amine-reactive fluorescent probe.
  • Maximum excitation/emission wavelength: 502/511 nm.
  • High stability.
  • Insensitive to polarity, pH, and solvent type.
  • Maintains stable fluorescence properties under various environmental conditions.
  • Can be used for synthesis of protease substrates, live cell imaging, protein labeling, and immunoassay.
Catalog No. 50-003-44140 Supplier Medchemexpress LLC Supplier No. HY1102101MG
BODIPY FL NHS ester (BODIPY FL, SE) is a cell membrane-penetrable amine-reactive fluorescent probe. The maximum excitation/emission wavelength of the BODIPY-FL NHS ester are 502/511 nm, respectively. It has high stability and is insensitive to the polarity, pH and type of solvent, and can maintain stable fluorescence properties under different environmental conditions. It can be used for the synthesis of protease substrates, live cell imaging, protein labeling and immunoassay.
  • Cell membrane-penetrable amine-reactive fluorescent probe.
  • Maximum excitation/emission wavelength: 502/511 nm.
  • High stability.
  • Insensitive to polarity, pH, and solvent type.
  • Maintains stable fluorescence properties under various environmental conditions.
  • Can be used for synthesis of protease substrates, live cell imaging, protein labeling, and immunoassay.