Learn More
Medchemexpress LLC BODIPY-FL NHS ester | 146616-66-2 | 99.8% | 389.16 | 50 MG
SDP
Catalog No. 5000343673
Encompass_Preferred
BODIPY FL NHS ester is a cell membrane-penetrable amine-reactive fluorescent probe with maximum excitation/emission wavelengths of 502/511 nm. It demonstrates high stability and is unaffected by polarity, pH, or solvent type, maintaining stable fluorescence across various environmental conditions.
  • Can be utilized for the synthesis of protease substrates.
  • Suitable for live cell imaging.
  • Useful for protein labeling.
  • Applicable in immunoassay.
Catalog No. 50-003-43673 Supplier Medchemexpress LLC Supplier No. HY11021010MG
BODIPY FL NHS ester is a cell membrane-penetrable amine-reactive fluorescent probe with maximum excitation/emission wavelengths of 502/511 nm. It demonstrates high stability and is unaffected by polarity, pH, or solvent type, maintaining stable fluorescence across various environmental conditions.
  • Can be utilized for the synthesis of protease substrates.
  • Suitable for live cell imaging.
  • Useful for protein labeling.
  • Applicable in immunoassay.