Learn More
Medchemexpress LLC BODIPY-FL NHS ester | 146616-66-2 | MFCD00467385 | 99.8% | 389.16 g·mol⁻¹ | C18H18BF2N3O4 | 5 MG
SDP
Catalog No. 5000263482
Encompass_Preferred
BODIPY-FL NHS ester is an amine-reactive, membrane-permeable fluorescent dye with excitation around 502 nm and emission around 510-511 nm, used to label proteins, peptides, and other primary-amine-containing molecules for research applications. It offers high photostability and maintains fluorescence across a range of solvent polarities and pH conditions.
  • Amine-reactive NHS ester for efficient conjugation to primary amines.
  • Excitation ~502 nm and emission ~510-511 nm for green fluorescence detection.
  • High photostability and stable fluorescence across pH and solvent conditions.
  • High purity suitable for labeling, imaging, and assay development.
  • Available as a solid in multiple sizes for flexible experimental use.
Catalog No. 50-002-63482 Supplier Medchemexpress LLC Supplier No. HY1102105MG
BODIPY-FL NHS ester is an amine-reactive, membrane-permeable fluorescent dye with excitation around 502 nm and emission around 510-511 nm, used to label proteins, peptides, and other primary-amine-containing molecules for research applications. It offers high photostability and maintains fluorescence across a range of solvent polarities and pH conditions.
  • Amine-reactive NHS ester for efficient conjugation to primary amines.
  • Excitation ~502 nm and emission ~510-511 nm for green fluorescence detection.
  • High photostability and stable fluorescence across pH and solvent conditions.
  • High purity suitable for labeling, imaging, and assay development.
  • Available as a solid in multiple sizes for flexible experimental use.