Learn More
Lumiprobe BDP FL NHS ester, 146616-66-2, 5mg
Catalog No. NC0656839
Encompass_Preferred
Molecular formula: C18H18BF2N3O4, Molecular weight: 389.16, Appearance: orange solid, Solubility: Good in organic solvents (DMF, DMSO, dichlromethane), limited in water; Very bright and photostable green emitting fluorescent dye; an analog of BODIPY FL. Amine-reactive dye for the labeling of proteins/peptides and amino-oligonucleotides.
Catalog No. NC0656839 Supplier Lumiprobe Supplier No. 21420
Molecular formula: C18H18BF2N3O4, Molecular weight: 389.16, Appearance: orange solid, Solubility: Good in organic solvents (DMF, DMSO, dichlromethane), limited in water; Very bright and photostable green emitting fluorescent dye; an analog of BODIPY FL. Amine-reactive dye for the labeling of proteins/peptides and amino-oligonucleotides.