Learn More
Medchemexpress LLC BODIPY-FL NHS ester (BODIPY FL,SE) | 146616-66-2 | 99.8% | 389.16 | 10 MG
SDP
Catalog No. 5000344230
Encompass_Preferred
BODIPY FL NHS ester (BODIPY FL, SE) is a cell membrane-penetrable amine-reactive fluorescent probe. Its maximum excitation/emission wavelengths are 502/511 nm, respectively. It exhibits high stability and is insensitive to polarity, pH, and solvent type, maintaining stable fluorescence properties across various environmental conditions. This ester can be utilized for synthesizing protease substrates, live cell imaging, protein labeling, and immunoassay.
  • Cell membrane-penetrable
  • Amine-reactive fluorescent probe
  • Maximum excitation/emission wavelengths: 502/511 nm
  • High stability
  • Insensitive to polarity, pH, and solvent type
  • Maintains stable fluorescence properties under different environmental conditions
  • Can be used for the synthesis of protease substrates, live cell imaging, protein labeling, and immunoassay
Catalog No. 50-003-44230 Supplier Medchemexpress LLC Supplier No. HY110210100MG
BODIPY FL NHS ester (BODIPY FL, SE) is a cell membrane-penetrable amine-reactive fluorescent probe. Its maximum excitation/emission wavelengths are 502/511 nm, respectively. It exhibits high stability and is insensitive to polarity, pH, and solvent type, maintaining stable fluorescence properties across various environmental conditions. This ester can be utilized for synthesizing protease substrates, live cell imaging, protein labeling, and immunoassay.
  • Cell membrane-penetrable
  • Amine-reactive fluorescent probe
  • Maximum excitation/emission wavelengths: 502/511 nm
  • High stability
  • Insensitive to polarity, pH, and solvent type
  • Maintains stable fluorescence properties under different environmental conditions
  • Can be used for the synthesis of protease substrates, live cell imaging, protein labeling, and immunoassay