Learn More
Echelon Biosciences Research Labs BODIPY FL-PI(3,4)P2 100 ug
SDP
Catalog No. 50720090
Encompass
(BODIPY FL Phosphatidylinositol 3,4-bisphosphate) is a water soluble analog of PI(3,4)P2 labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) is produced in stimulated cells by the action of phosphatidylinositol 3-kinases (PI3Ks) and/or lipid phosphatases (SHIP, TPTE, etc.). PI(3,4)P2 binds the pleckstrin homology (PH)-domain of AKT/PKB leading to cell survival and it also has an important role in podosome formation near focal adhesions. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-090 Supplier Echelon Biosciences Research Labs Supplier No. C34F6100UG
Add to Cart
Add to Cart
(BODIPY FL Phosphatidylinositol 3,4-bisphosphate) is a water soluble analog of PI(3,4)P2 labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) is produced in stimulated cells by the action of phosphatidylinositol 3-kinases (PI3Ks) and/or lipid phosphatases (SHIP, TPTE, etc.). PI(3,4)P2 binds the pleckstrin homology (PH)-domain of AKT/PKB leading to cell survival and it also has an important role in podosome formation near focal adhesions. This item is non returnable