Learn More
Echelon Biosciences Research Labs BODIPY FL-PI(3,5)P2 50 ug
SDP
Catalog No. 50720105
Encompass
BODIPY FL PI(3,5)P2 (BODIPY FL Phosphatidylinositol 3,5-bisphosphate) is a water soluble analog of PI(3,5)P2 labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,5-bisphosphate is in very low abundance and is required for retrograde membrane trafficking from lysosomal and late endosomal compartments to the Golgi and is involved in autophagy. PI(3,5)P2 levels are controlled by the 5-kinase PIKfyve and the 5-phosphatase Fig4/Sac3 and disregulation is linked to several human neuropathies (eg Chacot-Marie-Tooth disease). This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-105 Supplier Echelon Biosciences Research Labs Supplier No. C35F650UG
BODIPY FL PI(3,5)P2 (BODIPY FL Phosphatidylinositol 3,5-bisphosphate) is a water soluble analog of PI(3,5)P2 labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,5-bisphosphate is in very low abundance and is required for retrograde membrane trafficking from lysosomal and late endosomal compartments to the Golgi and is involved in autophagy. PI(3,5)P2 levels are controlled by the 5-kinase PIKfyve and the 5-phosphatase Fig4/Sac3 and disregulation is linked to several human neuropathies (eg Chacot-Marie-Tooth disease). This item is non returnable