Learn More
Echelon Biosciences Research Labs BODIPY FL Phosphatidylinositol 50 ug
SDP
Catalog No. 50720030
Encompass
BODIPY FL Phosphatidylinositol is a water soluble analog of Phosphatidylinositol (PI) labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol (PI or PtdIns) is biosynthesized from CDP-diacylglycerol and inositol by phosphatidylinositol synthase. PI is the building block for phosphoinositide polyphosphates through phosphorylation by PI-kinases and is a substrate for PI-PLC yielding diacylgelycerol. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-030 Supplier Echelon Biosciences Research Labs Supplier No. C00F650UG
BODIPY FL Phosphatidylinositol is a water soluble analog of Phosphatidylinositol (PI) labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol (PI or PtdIns) is biosynthesized from CDP-diacylglycerol and inositol by phosphatidylinositol synthase. PI is the building block for phosphoinositide polyphosphates through phosphorylation by PI-kinases and is a substrate for PI-PLC yielding diacylgelycerol. This item is non returnable