Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BODIPY™ FL, STP Ester, Sodium Salt

Catalog No. B10006
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10006 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B10006 Supplier Invitrogen™ Supplier No. B10006
Only null left
Add to Cart
Add to Cart

BODIPY™ FL, STP ester, sodium salt

The amine-reactive BODIPY™ FL STP ester produces electronically neutral dye conjugates that are spectrally similar to the negatively charged fluorescein dye. This dye's lack of ionic charge results in minimal effects on the isoelectric points of standard proteins conjugated with this fluorophore. The small size and relatively long excited-state lifetime of BODIPY™ FL dye has proven useful for studying ligand-receptor interaction by fluorescence polarization. In addition, BODIPY™ FL dye has little or no spectral overlap with longer-wavelength dyes such as tetramethylrhodamine and Texas Red™ dye, making it a useful green fluorophore for multicolor applications. The STP ester has higher water solubility than simple succinimidyl esters; the resulting dye conjugate is the same.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type FL STP Ester
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, STP Ester
Label or Dye BODIPY™ FL
Label Type BODIPY Dyes
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.