Learn More
Cayman Chemical BODIPY FL succinimide ester, Purity: ≥95%, CAS: 146616-66-2, 1 mg
SDP
Catalog No. 502992643
Encompass_Preferred
An amine-reactive fluorescent probe; ex/em = 502/511 nm; has been used in the synthesis of protease substrates that are non-fluorescent until unquenched following proteolytic cleavage; has been used in the synthesis of a fluorescent verapamil derivative and of fluorescent inhibitors of cholesterol absorption
This item is not returnable. View return policy
Catalog No. 50-299-2643 Supplier Cayman Chemical Supplier No. 295081 MG
An amine-reactive fluorescent probe; ex/em = 502/511 nm; has been used in the synthesis of protease substrates that are non-fluorescent until unquenched following proteolytic cleavage; has been used in the synthesis of a fluorescent verapamil derivative and of fluorescent inhibitors of cholesterol absorption