Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BODIPY-Palmitate | 1246809-50-6 | 97.3% | 487.45 | 1 MG
SDP

Catalog No. 5000393486
Encompass_Preferred

BODIPY-Palmitate is a fluorescently labeled palmitic acid, with excitation/emission maxima at 488/508 nm. It can be used to detect fatty acid uptake and metabolism.

  • Fluorescently labeled palmitic acid
  • Excitation/emission maxima at 488/508 nm
  • Detects fatty acid uptake and metabolism
  • Appearance: solid
  • Color: yellow to orange

Catalog No. 50-003-93486 Supplier Medchemexpress LLC Supplier No. HY1683751MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BODIPY-Palmitate is a fluorescently labeled palmitic acid, with excitation/emission maxima at 488/508 nm. It can be used to detect fatty acid uptake and metabolism.

  • Fluorescently labeled palmitic acid
  • Excitation/emission maxima at 488/508 nm
  • Detects fatty acid uptake and metabolism
  • Appearance: solid
  • Color: yellow to orange

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.