Learn More
Medchemexpress LLC BODIPY-palmitate | 1246809-50-6 | ≥98.0% | 487.45 g/mol | C28H42BF2N2O2- | 10 MG
SDP
Catalog No. 5000294985
Encompass_Preferred
BODIPY-Palmitate is a fluorescently labeled palmitic acid probe used to monitor fatty acid uptake and metabolism in cells. It has excitation/emission maxima at 488/508 nm and is provided in small milligram quantities for research use in fluorescence-based assays.
  • Excitation/emission maxima of 488/508 nm.
  • Suitable for detecting fatty acid uptake and metabolism in cultured cells.
  • Available in small milligram quantities for research applications.
  • High purity (≥98.0%) for consistent fluorescence performance.
  • Compatible with fluorescence microscopy and flow cytometry workflows.
Catalog No. 50-002-94985 Supplier Medchemexpress LLC Supplier No. HY16837510MG
BODIPY-Palmitate is a fluorescently labeled palmitic acid probe used to monitor fatty acid uptake and metabolism in cells. It has excitation/emission maxima at 488/508 nm and is provided in small milligram quantities for research use in fluorescence-based assays.
  • Excitation/emission maxima of 488/508 nm.
  • Suitable for detecting fatty acid uptake and metabolism in cultured cells.
  • Available in small milligram quantities for research applications.
  • High purity (≥98.0%) for consistent fluorescence performance.
  • Compatible with fluorescence microscopy and flow cytometry workflows.