Learn More
Cayman Chemical BODIPY-Palmitate, Purity: ≥98%, CAS: 1246809-50-6, 1 mg
SDP
Catalog No. 502937014
Encompass_Preferred
Fluorescently tagged palmitic acid; ex/em = 488/508 nm, respectively; has been used to monitor fatty acid uptake and metabolism in cultured cells
Catalog No. 50-293-7014 Supplier Cayman Chemical Supplier No. 267491 MG
Fluorescently tagged palmitic acid; ex/em = 488/508 nm, respectively; has been used to monitor fatty acid uptake and metabolism in cultured cells