Learn More
Echelon Biosciences Research Labs BODIPY TMR-PI(3,4)P2 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs C34M6100UG
Encompass
BODIPY TMR Phosphatidylinositol 3,4-bisphosphate is a water soluble analog of PI(3,4)P2 labeled with the orange fluorophore, Bodipy-TMR, at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,4-bisphosphate (PI(3,4)P2) is produced in stimulated cells by the action of phosphatidylinositol 3-kinases (PI3Ks) and/or lipid phosphatases (SHIP, TPTE, etc.). PI(3,4)P2 binds the pleckstrin homology (PH)-domain of AKT/PKB leading to cell survival and it also has an important role in podosome formation near focal adhesions. This item is non returnable
Catalog No. 50-720-094