Learn More
Echelon Biosciences Research Labs BODIPY TMR-PTDINS(3)P 100 ug
SDP
Catalog No. 50720049
Encompass
BODIPY TMR PI(3)P (BODIPY TMR Phosphatidylinositol 3-phosphate) is a water soluble analog of PI(3)P labeled with the orange fluorophore, Bodipy-TMR, at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3-phosphate (PI(3)P) is enriched in early endosomes having roles in endosome fusion and receptor sorting and internalization in multivesicular bodies. PI(3)P has also been found at the plasma membrane and is involved in the translocation of the glucose transport protein GLUT4. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-049 Supplier Echelon Biosciences Research Labs Supplier No. C03M6100UG
BODIPY TMR PI(3)P (BODIPY TMR Phosphatidylinositol 3-phosphate) is a water soluble analog of PI(3)P labeled with the orange fluorophore, Bodipy-TMR, at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3-phosphate (PI(3)P) is enriched in early endosomes having roles in endosome fusion and receptor sorting and internalization in multivesicular bodies. PI(3)P has also been found at the plasma membrane and is involved in the translocation of the glucose transport protein GLUT4. This item is non returnable