Learn More
Echelon Biosciences Research Labs BODIPY TMR-PI(5)P 100 ug
SDP
Catalog No. 50720077
Encompass
BODIPY TMR PI(5)P (BODIPY TMR Phosphatidylinositol 5-phosphate) is a water soluble analog of PI(5)P labeled with the orange fluorophore, Bodipy-TMR, at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 5-phosphate (PI(5)P) is a rare lipid found in the nucleus that can bind the PHD finger of the nuclear adapter protein ING2. PI(5)P is phosphorylated to PI(4,5)P2 by Type II PIP kinases. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-077 Supplier Echelon Biosciences Research Labs Supplier No. C05M6100UG
BODIPY TMR PI(5)P (BODIPY TMR Phosphatidylinositol 5-phosphate) is a water soluble analog of PI(5)P labeled with the orange fluorophore, Bodipy-TMR, at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 5-phosphate (PI(5)P) is a rare lipid found in the nucleus that can bind the PHD finger of the nuclear adapter protein ING2. PI(5)P is phosphorylated to PI(4,5)P2 by Type II PIP kinases. This item is non returnable