Learn More
Echelon Biosciences Research Labs BODIPY TMR Phosphatidylinositol 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs C00M650UG
Encompass
BODIPY TMR Phosphatidylinositol is a water soluble analog of Phosphatidylinositol (PI) labeled with the orange fluorophore, Bodipy-TMR, at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol (PI or PtdIns) is biosynthesized from CDP-diacylglycerol and inositol by phosphatidylinositol synthase. PI is the building block for phosphoinositide polyphosphates through phosphorylation by PI-kinases and is a substrate for PI-PLC yielding diacylgelycerol. This item is non returnable
Catalog No. 50-720-034