Learn More
Medchemexpress LLC Bodipy-TS (Thiol-green 2) | 1808174-52-8 | 97.1% | 434.33 | C20H21BF2N2O2S2 | 10 MG
SDP
Catalog No. 5000297254
Encompass_Preferred
BODIPY-TS (Thiol-green 2) is a thiol-specific, turn-on fluorescent probe based on a BODIPY dye scaffold with a thiosulfonate recognition unit. It is intended for rapid and selective detection of thiol-containing species in research applications.
  • Thiol-specific turn-on fluorescence response.
  • Fast response and high selectivity for thiol compounds.
  • Offered in small-scale laboratory pack sizes, including 10 MG.
  • High reported purity (97.1%).
  • Suitable for research applications.
Catalog No. 50-002-97254 Supplier Medchemexpress LLC Supplier No. HYD126210MG
BODIPY-TS (Thiol-green 2) is a thiol-specific, turn-on fluorescent probe based on a BODIPY dye scaffold with a thiosulfonate recognition unit. It is intended for rapid and selective detection of thiol-containing species in research applications.
  • Thiol-specific turn-on fluorescence response.
  • Fast response and high selectivity for thiol compounds.
  • Offered in small-scale laboratory pack sizes, including 10 MG.
  • High reported purity (97.1%).
  • Suitable for research applications.