Learn More
Medchemexpress LLC BODIPY-X-Alkyne | 1173281-82-7 | 96.0% | 434.33 | 50 MG
SDP
Catalog No. 5000429992
Encompass_Preferred
BODIPY-X-Alkyne is an alkyne-modified BODIPY derivative with excitation at 526 nm and emission at 540 nm. It can react with azides via a copper-catalyzed click reaction.
  • Alkyne-modified BODIPY derivative
  • Excitation (Ex) = 525 nm
  • Emission (Em) = 540 nm
  • Reacts with azides via a copper-catalyzed click reaction
Catalog No. 50-004-29992 Supplier Medchemexpress LLC Supplier No. HYD158850MG
BODIPY-X-Alkyne is an alkyne-modified BODIPY derivative with excitation at 526 nm and emission at 540 nm. It can react with azides via a copper-catalyzed click reaction.
  • Alkyne-modified BODIPY derivative
  • Excitation (Ex) = 525 nm
  • Emission (Em) = 540 nm
  • Reacts with azides via a copper-catalyzed click reaction