Learn More
Mopec Body Bag, COVID-19
SDP
Catalog No. p-9034001 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Standard-duty Body Bag
Includes:
No Handles
Catalog No. Product Type Includes
22-445-151 Standard-duty Body Bag No Handles
1 options
Catalog No. 22-445-151 Supplier Mopec Supplier No. BE166
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Mopec has partnered with multiple vinyl and polyethylene manufacturers to meet the demand for body bags. These new partnerships will enable us to meet the large quantities needed across the country.
Emergency response organizations, Medical Examiner offices, hospitals, and morgues across the country are quickly depleting stocks of standard and heavy duty body bags. Many have resorted to using thin lightweight or economy bags while increasing to using two or three per decedent to ensure durability and safety. This sudden demand has left the few body bag manufactures and suppliers in the country with the inability to keep up with the demand. Even with adding more employees and multiple shifts, body bag manufacturers are reporting backlogs of three to six months, or longer. Recognizing this need, Mopec began pursuing manufacturers across the continent to assist in the effort. This bag will come in whatever colors are available and in stock.
Specifications
Description Leak Proof: Seams are 100% RF heat welded/sealed
Closure Type Center Zipper
Color White
Includes No Handles
Material PVC
Thickness (English) 7 to 12 mil
Dimensions (L x W) 94 x 36 in.
Weight (English) 200+ pounds
Product Type Standard-duty Body Bag