Learn More
DSI LLC Body, Cheminert mixing tee 1/4"-28, for 1/8" tubing, 1.5 mm bore, PEEK
Catalog No. 502255017
Encompass_Preferred
Body, Cheminert mixing tee 1/4"-28, for 1/8" tubing, 1.5 mm bore, PEEK
Catalog No. 50-225-5017 Supplier DSI LLC Supplier No. JRBCM2XPK
Body, Cheminert mixing tee 1/4"-28, for 1/8" tubing, 1.5 mm bore, PEEK