Learn More
MSC Honeywell Miller Vest-Style Full Body Harness, Elastomer, L/XL, 1 ea
Catalog No. 5000507122
Encompass
This full body, vest-style harness is engineered for safety and comfort in construction and confined space applications. Constructed with specially-formulated elastomer webbing, the harness provides a flexible fit that improves mobility and productivity. It includes integrated back and leg padding for enhanced comfort during extended wear.
  • Specially-formulated elastomer webbing stretches to provide increased comfort and safety
  • Features integrated back and leg pads
  • Equipped with a sliding back D-ring for support
  • Includes mating buckle chest and leg straps with friction buckle shoulder straps
  • Designed with pull-free lanyard rings and belt loops
  • Meets or exceeds ANSI, CSA, and OSHA standards
  • Capacity of 400 lbs
Catalog No. 50-005-07122 Supplier MSC Supplier No. 91515270
This full body, vest-style harness is engineered for safety and comfort in construction and confined space applications. Constructed with specially-formulated elastomer webbing, the harness provides a flexible fit that improves mobility and productivity. It includes integrated back and leg padding for enhanced comfort during extended wear.
  • Specially-formulated elastomer webbing stretches to provide increased comfort and safety
  • Features integrated back and leg pads
  • Equipped with a sliding back D-ring for support
  • Includes mating buckle chest and leg straps with friction buckle shoulder straps
  • Designed with pull-free lanyard rings and belt loops
  • Meets or exceeds ANSI, CSA, and OSHA standards
  • Capacity of 400 lbs