Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPX3301239 Quick-disconnect body, polysulfone, 3/4"" sanitary connector; MPX
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3160426
Encompass
These couplings allow you to integrate disposables with your existing stainless steel processing equipment. Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps. Choose from stainless steel or polysulfone inserts to mate to polysulfone bodies. All models feature USP Class VI platinum-cured silicone gaskets. Mix and match these bodies and inserts with the polysulfone quick-disconnect coupling elements on page 509 for greater flexibility in your process system.
Catalog No. 50-627-923