Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC BODYHAIR SHAMPOO

Catalog No. 502819151
Encompass

This liquid soap is designed for head-to-toe use, offering a viscous, green-yellow appearance and a pleasant herbal scent. It is supplied in 1 Qt bottles.

  • Formulated for hair and body cleansing
  • Viscous liquid consistency
  • Pleasant herbal scent
  • Supplied in 1 Qt bottles
  • PH range of 6.5-8.5
  • Density of 1.02 g/cm³
  • Water soluble

Catalog No. 50-281-9151 Supplier MSC Supplier No. 15607021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This liquid soap is designed for head-to-toe use, offering a viscous, green-yellow appearance and a pleasant herbal scent. It is supplied in 1 Qt bottles.

  • Formulated for hair and body cleansing
  • Viscous liquid consistency
  • Pleasant herbal scent
  • Supplied in 1 Qt bottles
  • PH range of 6.5-8.5
  • Density of 1.02 g/cm³
  • Water soluble

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.