Learn More
Boekel Scientific™ Microplate Shaker Platform
GREENER_CHOICESDP
Catalog No. 15600322 Shop All Boekel Scientific Products
Change view
Click to view available options
Holds:
4 Microplates
Catalog No. Holds
15-600-322 4 Microplates
1 options
Catalog No. 15-600-322 Supplier Boekel Scientific™ Supplier No. 270390
Add to Cart
Add to Cart
Need to upgrade your Boekel Microplate Shaker to hold more plates? This 4 plate microplate tray is adaptable to the Boekel 2 Plate Microplate Shaker.
  • interchangeable tray for holding microplates
  • Simple screw attachments allow quick and easy fitting of any standard depth 96- or 384-well plates, holding the plates securely in place
Specifications
Holds 4 Microplates
For Use With (Equipment) Boekel Scientific Microplate Shaker
No. of Positions 4