Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boekel Scientific™ Small Desiccator
GREENER_CHOICESDP

Catalog No. 0864540 Shop All Boekel Scientific Products
Change view
Click to view available options
Cabinet Style:
Front-loading
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cabinet Style
08-645-40 Front-loading
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

No vacuum or pressure should be applied.

Catalog No. 08-645-40 Supplier Boekel Scientific™ Supplier No. 1340
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

No vacuum or pressure should be applied.

Small Desiccator for storing reagents, IR cells and soil samples.

  • Durable, hand-crafted construction
  • Fabricated from stainless steel and glass
  • With seal integrity
  • Rigid door full-length door hinges
  • One-piece neoprene door gasket
  • Adjustable shelves with 1/2 in. increments
  • Automatic desiccant regeneration available
  • For Drying biological specimens, Cooling crucibles, Storage sludge samples

Specifications

Cabinet Style Front-loading
Material Stainless Steel, Glass
Product Type Nonvacuum Desiccator
For Use With (Application) Storing reagents or soil samples, drying biological specimens, storage sludge samples, cooling down from 40° to 150°C
Includes Two cement board shelves and desiccator tray
Size Small
Warranty 2 Years
Dimensions (L x W x H) 12.25 x 12.5 x 12.25 in. (31.1 x 31.7 x 31 cm)
Length (English) 12.25 in.
Length (English) Chamber 10 in.
Length (Metric) 31.1 cm
Length (Metric) Chamber 25.4 cm
Width (English) 12.5 in.
Width (English) Chamber 11.5 in.
Width (Metric) 31.7 cm
Width (Metric) Chamber 29.2 cm
Height (English) 12.25 in.
Height (English) Chamber 12 in.
Height (Metric) 31 cm
Height (Metric) Chamber 30.5 cm
Depth (English) 12.25 in.
Depth (Metric) 31.1 cm
Weight (English) 23 lb.
Weight (Metric) 10.4 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.