Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boeravinone E | 137787-00-9 | 1 MG
SDP

Catalog No. 5000421224
Encompass_Preferred

Boeravinone E is a natural product isolated from *Boerhaavia diffusa L*, classified as a flavonoid, isoflavone, phenol, and polyphenol. It exhibits spasmolytic activity and is intended for research use only.

  • Purity of 99.73%
  • Molecular weight of 328.27
  • Chemical formula: C17H12O7
  • Appears as a solid
  • Color is white to yellow
  • Classified as a flavonoid, isoflavone, phenol, and polyphenol
  • Initially sourced from *Boerhaavia diffusa L* plants
  • Ships at room temperature in continental US; may vary elsewhere
  • Store at 4°C, protected from light. In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-004-21224 Supplier Medchemexpress LLC Supplier No. HYN29481MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boeravinone E is a natural product isolated from *Boerhaavia diffusa L*, classified as a flavonoid, isoflavone, phenol, and polyphenol. It exhibits spasmolytic activity and is intended for research use only.

  • Purity of 99.73%
  • Molecular weight of 328.27
  • Chemical formula: C17H12O7
  • Appears as a solid
  • Color is white to yellow
  • Classified as a flavonoid, isoflavone, phenol, and polyphenol
  • Initially sourced from *Boerhaavia diffusa L* plants
  • Ships at room temperature in continental US; may vary elsewhere
  • Store at 4°C, protected from light. In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.