Learn More
Cayman Chemical BofutrelvIr 10mg
SDP
Catalog No. 502990779
Encompass_Preferred
An inhibitor of Mpro (IC50 = 0.053 µM in a TR-FRET assay); reduces viral yield in the culture supernatant of SARS-CoV-2-infected Vero E6 cells (EC50 = 0.53 µM); reduces viral RNA copy numbers in the same model from 1.85-50 µM
Catalog No. 50-299-0779 Supplier Cayman Chemical Supplier No. 3134410 MG
An inhibitor of Mpro (IC50 = 0.053 µM in a TR-FRET assay); reduces viral yield in the culture supernatant of SARS-CoV-2-infected Vero E6 cells (EC50 = 0.53 µM); reduces viral RNA copy numbers in the same model from 1.85-50 µM