Learn More
Medchemexpress LLC HY-12843 5mg Medchemexpress, Bohemine CAS:189232-42-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128435MG
Encompass_Preferred
Medchemexpress, HY-12843 5mg Bohemine CAS:189232-42-6 Bohemine is a purine analogue and is a synthetic and selective CDK inhibitor with IC50s of 4.6 μM, 83 μM, and 2.7 μM for Cdk2/cyclin E, Cdk2/cyclin A, and Cdk9/cyclin T1, respectively. Bohemine also inhibits ERK2 with an IC50 of 52 μM and has less inhibitory effect on CDK1, CDK4 and CDK6. Bohemine has a broad spectrum anti-cancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1168