Learn More
Electron Microscopy Sciences Bohler Bone Rongeur Double Action, Straight
SDP
Catalog No. 5030183
Encompass_Preferred
The jaw width is 3 mm and they are 6" long. Made from stainless steel with a polished finish.
Catalog No. 50-301-83 Supplier Electron Microscopy Sciences Supplier No. 77988S
The jaw width is 3 mm and they are 6" long. Made from stainless steel with a polished finish.