Learn More
Hach Company Boiler/Condens Conduct Sens
Catalog No. 5000207643 Shop All Hach Company Products
Encompass
Analog Boiler/Condensate Sensor-Cell Constant 0 5-Aluminum HDNominal Cell Constant (K) 0 5 3/4 inch male NPT threaded 316 stainless steel body Aluminum Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode
Catalog No. 50-002-07643 Supplier Hach Company Supplier No. 3444B8C
Analog Boiler/Condensate Sensor-Cell Constant 0 5-Aluminum HDNominal Cell Constant (K) 0 5 3/4 inch male NPT threaded 316 stainless steel body Aluminum Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable Titanium Electrode